Assay Details
| Gene Symbol: Eip63E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043116.3 | NP_001036581.1 | 9 - 10 | 2485 | 65 | |
| NM_001043117.3 | NP_001036582.1 | 10 - 11 | 1574 | 65 | ||
| NM_001043118.3 | NP_001036583.1 | 11 - 12 | 1879 | 65 | ||
| NM_001043119.3 | NP_001036584.1 | 12 - 13 | 1688 | 65 | ||
| NM_001043120.3 | NP_001036585.1 | 11 - 12 | 1907 | 65 | ||
| NM_001043121.2 | NP_001036586.1 | 9 - 10 | 1746 | 65 | ||
| NM_001169869.2 | NP_001163340.1 | 10 - 11 | 2260 | 65 | ||
| NM_001202113.2 | NP_001189042.1 | 10 - 11 | 1891 | 65 | ||
| NM_001274444.1 | NP_001261373.1 | 10 - 11 | 2524 | 65 | ||
| NM_079180.4 | NP_523904.1 | 10 - 11 | 2524 | 65 | ||
| NM_168033.3 | NP_728888.1 | 11 - 12 | 1930 | 65 | ||
| NM_168034.3 | NP_728889.1 | 11 - 12 | 1649 | 65 | ||
| NM_168035.2 | NP_728890.1 | 10 - 11 | 1868 | 65 | ||
| NM_168036.3 | NP_728891.1 | 10 - 11 | 1840 | 65 | ||
| GenBank mRNA | BT010064.1 | - | 9 - 10 | 1746 | 65 | |
| BT022123.1 | - | 1840 | 65 | |||
More Information
Set Membership: |
|

Eip63E
Genomic Map