Assay Details
| Gene Symbol: Eip63E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043117.3 | NP_001036582.1 | 4 - 5 | 443 | 91 | |
| NM_001043118.3 | NP_001036583.1 | 5 - 6 | 748 | 91 | ||
| NM_001043119.3 | NP_001036584.1 | 6 - 7 | 557 | 91 | ||
| NM_001043120.3 | NP_001036585.1 | 5 - 6 | 776 | 91 | ||
| NM_001169869.2 | NP_001163340.1 | 4 - 5 | 1129 | 91 | ||
| NM_001274444.1 | NP_001261373.1 | 4 - 5 | 1393 | 91 | ||
| NM_079180.4 | NP_523904.1 | 4 - 5 | 1393 | 91 | ||
| NM_168033.3 | NP_728888.1 | 5 - 6 | 799 | 91 | ||
| GenBank mRNA | AF152398.1 | - | 4 - 5 | 1458 | 91 | |
| AF152400.1 | - | 5 - 6 | 867 | 91 | ||
| AF152401.1 | - | 5 - 6 | 774 | 91 | ||
| AF152403.1 | - | 4 - 5 | 443 | 91 | ||
| AF152404.1 | - | 6 - 7 | 557 | 91 | ||
| AF152405.1 | - | 4 - 5 | 1148 | 91 | ||
| AF152406.1 | - | 5 - 6 | 744 | 91 | ||
More Information
Set Membership: |
|

Eip63E
Genomic Map