Assay Details
| Gene Symbol: Eip63E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043116.3 | NP_001036581.1 | 6 - 7 | 1835 | 68 | |
| NM_001043117.3 | NP_001036582.1 | 7 - 8 | 924 | 68 | ||
| NM_001043118.3 | NP_001036583.1 | 8 - 9 | 1229 | 68 | ||
| NM_001043119.3 | NP_001036584.1 | 9 - 10 | 1038 | 68 | ||
| NM_001043120.3 | NP_001036585.1 | 8 - 9 | 1257 | 68 | ||
| NM_001043121.2 | NP_001036586.1 | 6 - 7 | 1096 | 68 | ||
| NM_001169869.2 | NP_001163340.1 | 7 - 8 | 1610 | 68 | ||
| NM_001202113.2 | NP_001189042.1 | 7 - 8 | 1241 | 68 | ||
| NM_001274444.1 | NP_001261373.1 | 7 - 8 | 1874 | 68 | ||
| NM_079180.4 | NP_523904.1 | 7 - 8 | 1874 | 68 | ||
| NM_168033.3 | NP_728888.1 | 8 - 9 | 1280 | 68 | ||
| NM_168034.3 | NP_728889.1 | 8 - 9 | 999 | 68 | ||
| NM_168035.2 | NP_728890.1 | 7 - 8 | 1218 | 68 | ||
| NM_168036.3 | NP_728891.1 | 7 - 8 | 1190 | 68 | ||
| GenBank mRNA | BT010064.1 | - | 6 - 7 | 1096 | 68 | |
| BT022123.1 | - | 1190 | 68 | |||
More Information
Set Membership: |
|

Eip63E
Genomic Map