Assay Details
| Gene Symbol: Eip63E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043116.3 | NP_001036581.1 | 8 - 9 | 2304 | 58 | |
| NM_001043117.3 | NP_001036582.1 | 9 - 10 | 1393 | 58 | ||
| NM_001043118.3 | NP_001036583.1 | 10 - 11 | 1698 | 58 | ||
| NM_001043119.3 | NP_001036584.1 | 11 - 12 | 1507 | 58 | ||
| NM_001043120.3 | NP_001036585.1 | 10 - 11 | 1726 | 58 | ||
| NM_001043121.2 | NP_001036586.1 | 8 - 9 | 1565 | 58 | ||
| NM_001169869.2 | NP_001163340.1 | 9 - 10 | 2079 | 58 | ||
| NM_001202113.2 | NP_001189042.1 | 9 - 10 | 1710 | 58 | ||
| NM_001274444.1 | NP_001261373.1 | 9 - 10 | 2343 | 58 | ||
| NM_079180.4 | NP_523904.1 | 9 - 10 | 2343 | 58 | ||
| NM_168033.3 | NP_728888.1 | 10 - 11 | 1749 | 58 | ||
| NM_168034.3 | NP_728889.1 | 10 - 11 | 1468 | 58 | ||
| NM_168035.2 | NP_728890.1 | 9 - 10 | 1687 | 58 | ||
| NM_168036.3 | NP_728891.1 | 9 - 10 | 1659 | 58 | ||
| GenBank mRNA | AF152398.1 | - | 9 - 10 | 2408 | 58 | |
| AF152399.1 | - | 8 - 9 | 2369 | 58 | ||
| AF152400.1 | - | 10 - 11 | 1817 | 58 | ||
| AF152401.1 | - | 10 - 11 | 1724 | 58 | ||
| AF152402.1 | - | 9 - 10 | 1685 | 58 | ||
| AF152403.1 | - | 9 - 10 | 1393 | 58 | ||
| AF152404.1 | - | 11 - 12 | 1507 | 58 | ||
| AF152405.1 | - | 9 - 10 | 2098 | 58 | ||
| AF152406.1 | - | 10 - 11 | 1694 | 58 | ||
| BT010064.1 | - | 8 - 9 | 1565 | 58 | ||
| BT022123.1 | - | 1659 | 58 | |||
More Information
Set Membership: |
|

Eip63E
Genomic Map