Assay Details
| Gene Symbol: ImpL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274480.1 | NP_001261409.1 | 2 - 3 | 1507 | 90 | |
| NM_079197.5 | NP_523921.3 | 2 - 3 | 1023 | 90 | ||
| NM_168069.2 | NP_728960.1 | 2 - 3 | 1507 | 90 | ||
| NM_168070.2 | NP_728961.2 | 2 - 3 | 845 | 90 | ||
| GenBank mRNA | AY060476.1 | - | 2 - 3 | 1475 | 90 | |
| BT012298.1 | - | 2 - 3 | 845 | 90 | ||
| BT012299.1 | - | 2 - 3 | 1023 | 90 | ||
More Information
Set Membership: |
|

ImpL2
Genomic Map