Assay Details
| Gene Symbol: Dhc64C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274498.1 | NP_001261427.1 | 1 - 2 | 358 | 102 | |
| NM_001274499.1 | NP_001261428.1 | 1 - 2 | 358 | 102 | ||
| NM_001274500.1 | NP_001261429.1 | 1 - 2 | 358 | 102 | ||
| NM_001274501.1 | NP_001261430.1 | 1 - 2 | 358 | 102 | ||
| NM_001274502.1 | NP_001261431.1 | 1 - 2 | 358 | 102 | ||
| NM_001274503.1 | NP_001261432.1 | 1 - 2 | 358 | 102 | ||
| NM_079205.4 | NP_523929.2 | 1 - 2 | 358 | 102 | ||
| NM_168103.3 | NP_729034.2 | 1 - 2 | 358 | 102 | ||
More Information
Set Membership: |
|

Dhc64C
Genomic Map