Assay Details
| Gene Symbol: Con | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259706.2 | NP_001246635.1 | 7 - 8 | 2241 | 97 | |
| NM_079206.5 | NP_523930.3 | 4 - 5 | 1710 | 97 | ||
| GenBank mRNA | BT015228.1 | - | 4 - 5 | 1706 | 97 | |
More Information
Set Membership: |
|

Con
Genomic Map