Assay Details
| Gene Symbol: S6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274521.1 | NP_001261450.1 | 3 - 4 | 814 | 123 | |
| NM_079217.3 | NP_523941.2 | 3 - 4 | 814 | 123 | ||
| GenBank mRNA | BT133257.1 | - | 3 - 4 | 812 | 123 | |
More Information
Set Membership: |
|

S6k
Genomic Map