Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104070.2 | NP_001097540.1 | 10 - 11 | 2575 | 72 | |
| NM_001259732.1 | NP_001246661.1 | 6 - 7 | 1855 | 72 | ||
| NM_079241.4 | NP_523965.2 | 11 - 12 | 4304 | 72 | ||
| NM_168241.3 | NP_729306.1 | 10 - 11 | 2930 | 72 | ||
| NM_168242.3 | NP_729307.1 | 9 - 10 | 2568 | 72 | ||
| NM_168243.3 | NP_729308.1 | 10 - 11 | 3942 | 72 | ||
| GenBank mRNA | AF136492.1 | - | 10 - 11 | 2882 | 72 | |
| BT120091.1 | - | 6 - 7 | 1855 | 72 | ||
| BT150408.1 | - | 10 - 11 | 2898 | 72 | ||
More Information
Set Membership: |
|

pbl
Genomic Map