Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104070.2 | NP_001097540.1 | 4 - 5 | 642 | 60 | |
| NM_079241.4 | NP_523965.2 | 4 - 5 | 997 | 60 | ||
| NM_168241.3 | NP_729306.1 | 4 - 5 | 997 | 60 | ||
| NM_168242.3 | NP_729307.1 | 3 - 4 | 635 | 60 | ||
| NM_168243.3 | NP_729308.1 | 3 - 4 | 635 | 60 | ||
| GenBank mRNA | AF136492.1 | - | 4 - 5 | 949 | 60 | |
| BT150408.1 | - | 4 - 5 | 965 | 60 | ||
More Information
Set Membership: |
|

pbl
Genomic Map