Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079241.4 | NP_523965.2 | 6 - 7 | 2622 | 67 | |
| NM_168243.3 | NP_729308.1 | 5 - 6 | 2260 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pbl| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079241.4 | NP_523965.2 | 6 - 7 | 2622 | 67 | |
| NM_168243.3 | NP_729308.1 | 5 - 6 | 2260 | 67 | ||
Set Membership: |
|