Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104070.2 | NP_001097540.1 | 6 - 7 | 1020 | 65 | |
| NM_001259732.1 | NP_001246661.1 | 2 - 3 | 300 | 65 | ||
| NM_079241.4 | NP_523965.2 | 7 - 8 | 2749 | 65 | ||
| NM_168241.3 | NP_729306.1 | 6 - 7 | 1375 | 65 | ||
| NM_168242.3 | NP_729307.1 | 5 - 6 | 1013 | 65 | ||
| NM_168243.3 | NP_729308.1 | 6 - 7 | 2387 | 65 | ||
| GenBank mRNA | BT120091.1 | - | 2 - 3 | 300 | 65 | |
| BT150408.1 | - | 6 - 7 | 1343 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT120091.1, NM_001259732.1 transcripts.
Set Membership: |
|

pbl
Genomic Map