Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104070.2 | NP_001097540.1 | 7 - 8 | 1368 | 69 | |
| NM_001259732.1 | NP_001246661.1 | 3 - 4 | 648 | 69 | ||
| NM_079241.4 | NP_523965.2 | 8 - 9 | 3097 | 69 | ||
| NM_168241.3 | NP_729306.1 | 7 - 8 | 1723 | 69 | ||
| NM_168242.3 | NP_729307.1 | 6 - 7 | 1361 | 69 | ||
| NM_168243.3 | NP_729308.1 | 7 - 8 | 2735 | 69 | ||
| GenBank mRNA | BT120091.1 | - | 3 - 4 | 648 | 69 | |
More Information
Set Membership: |
|

pbl
Genomic Map