Assay Details
| Gene Symbol: nmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274626.1 | NP_001261555.1 | 4 - 5 | 1019 | 71 | |
| NM_001370048.1 | NP_001356976.1 | 3 - 4 | 848 | 71 | ||
| NM_079243.4 | NP_523967.2 | 3 - 4 | 848 | 71 | ||
| NM_168248.3 | NP_729316.1 | 4 - 5 | 1019 | 71 | ||
| NM_168249.3 | NP_729317.1 | 3 - 4 | 1355 | 71 | ||
| NM_168250.3 | NP_729318.1 | 3 - 4 | 848 | 71 | ||
| NM_168251.3 | NP_729319.1 | 4 - 5 | 1019 | 71 | ||
| NM_168252.3 | NP_729320.1 | 3 - 4 | 1355 | 71 | ||
| GenBank mRNA | AY069602.1 | - | 3 - 4 | 1329 | 71 | |
More Information
Set Membership: |
|

nmo
Genomic Map