Assay Details
| Gene Symbol: nmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274626.1 | NP_001261555.1 | 7 - 8 | 1866 | 72 | |
| NM_001370048.1 | NP_001356976.1 | 6 - 7 | 1695 | 72 | ||
| NM_079243.4 | NP_523967.2 | 6 - 7 | 1695 | 72 | ||
| NM_168248.3 | NP_729316.1 | 7 - 8 | 1866 | 72 | ||
| NM_168249.3 | NP_729317.1 | 6 - 7 | 2202 | 72 | ||
| NM_168250.3 | NP_729318.1 | 6 - 7 | 1695 | 72 | ||
| NM_168251.3 | NP_729319.1 | 7 - 8 | 1866 | 72 | ||
| NM_168252.3 | NP_729320.1 | 6 - 7 | 2202 | 72 | ||
| GenBank mRNA | AY069602.1 | - | 6 - 7 | 2176 | 72 | |
More Information
Set Membership: |
|

nmo
Genomic Map