Assay Details
| Gene Symbol: Nmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079245.3 | NP_523969.1 | 1 - 2 | 310 | 64 | |
| GenBank mRNA | AF132556.1 | - | 1 - 2 | 291 | 64 | |
| AY118963.1 | - | 1 - 2 | 296 | 64 | ||
More Information
Set Membership: |
|

Nmt
Genomic Map