Assay Details
| Gene Symbol: DNApol-alpha50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079248.4 | NP_523972.2 | 2 - 3 | 353 | 93 | |
| GenBank mRNA | AY071217.1 | - | 2 - 3 | 347 | 93 | |
More Information
Set Membership: |
|

DNApol-alpha50
Genomic Map