Assay Details
| Gene Symbol: Prm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079258.3 | NP_523982.2 | 7 - 8 | 1777 | 68 | |
| NM_168290.3 | NP_729405.1 | 7 - 8 | 1777 | 68 | ||
| GenBank mRNA | BT044164.1 | - | 7 - 8 | 1781 | 68 | |
| X62590.1 | - | 1740 | 68 | |||
More Information
Set Membership: |
|

Prm
Genomic Map