Assay Details
| Gene Symbol: dally | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259756.2 | NP_001246685.1 | 2 - 3 | 1147 | 60 | |
| NM_079259.4 | NP_523983.1 | 2 - 3 | 1147 | 60 | ||
| GenBank mRNA | BT011348.1 | - | 2 - 3 | 1146 | 60 | |
| U31985.1 | - | 3 - 4 | 1107 | 60 | ||
More Information
Set Membership: |
|

dally
Genomic Map