Assay Details
| Gene Symbol: dally | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259756.2 | NP_001246685.1 | 5 - 6 | 1938 | 80 | |
| NM_079259.4 | NP_523983.1 | 5 - 6 | 1938 | 80 | ||
| GenBank mRNA | BT011348.1 | - | 5 - 6 | 1937 | 80 | |
More Information
Set Membership: |
|

dally
Genomic Map