Assay Details
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079260.3 | NP_523984.1 | 2 - 3 | 558 | 57 | |
| GenBank mRNA | AF124743.1 | - | 2 - 3 | 530 | 57 | |
| BT001526.1 | - | 2 - 3 | 565 | 57 | ||
More Information
Set Membership: |
|

Mcm7
Genomic Map