Assay Details
Gene Symbol: smg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144453.2 | NP_001137925.1 | 3 - 4 | 3242 | 128 | |
NM_079263.3 | NP_523987.1 | 3 - 4 | 3447 | 128 | ||
NM_168309.3 | NP_729441.1 | 3 - 4 | 3465 | 128 | ||
NM_168310.3 | NP_729442.1 | 4 - 5 | 3339 | 128 | ||
GenBank mRNA | AF132213.1 | - | 3 - 4 | 3265 | 128 | |
AF159852.1 | - | 3 - 4 | 3407 | 128 | ||
AY058607.1 | - | 3 - 4 | 3451 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
