Assay Details
| Gene Symbol: Dronc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079293.4 | NP_524017.1 | 1 - 2 | 856 | 55 | |
| GenBank mRNA | AF104357.1 | - | 1 - 2 | 816 | 55 | |
| AJ242796.1 | - | 1 - 2 | 815 | 55 | ||
| AY058747.1 | - | 1 - 2 | 892 | 55 | ||
More Information
Set Membership: |
|

Dronc
Genomic Map