Assay Details
| Gene Symbol: E(z) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144460.3 | NP_001137932.1 | 2 - 3 | 974 | 64 | |
| NM_001274753.1 | NP_001261682.1 | 2 - 3 | 974 | 64 | ||
| NM_079297.3 | NP_524021.2 | 3 - 4 | 1160 | 64 | ||
| GenBank mRNA | AY051785.1 | - | 2 - 3 | 923 | 64 | |
More Information
Set Membership: |
|

E(z)
Genomic Map