Assay Details
| Gene Symbol: CycA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079306.4 | NP_524030.2 | 1 - 2 | 707 | 97 | |
| GenBank mRNA | AY058712.1 | - | 1 - 2 | 705 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CycA| Gene Symbol: CycA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079306.4 | NP_524030.2 | 1 - 2 | 707 | 97 | |
| GenBank mRNA | AY058712.1 | - | 1 - 2 | 705 | 97 | |
Set Membership: |
|