Assay Details
| Gene Symbol: CycA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259795.2 | NP_001246724.1 | 5 - 6 | 1580 | 61 | |
| NM_079306.4 | NP_524030.2 | 5 - 6 | 1583 | 61 | ||
| GenBank mRNA | AY058712.1 | - | 5 - 6 | 1581 | 61 | |
More Information
Set Membership: |
|

CycA
Genomic Map