Assay Details
| Gene Symbol: byn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274801.1 | NP_001261730.1 | 4 - 5 | 936 | 77 | |
| NM_079307.2 | NP_524031.2 | 4 - 5 | 936 | 77 | ||
More Information
Set Membership: |
|

byn
Genomic Map