Assay Details
| Gene Symbol: toe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274833.1 | NP_001261762.1 | 3 - 4 | 994 | 64 | |
| NM_079317.3 | NP_524041.2 | 3 - 4 | 994 | 64 | ||
| GenBank mRNA | AY069177.1 | - | 3 - 4 | 994 | 64 | |
More Information
Set Membership: |
|

toe
Genomic Map