Assay Details
| Gene Symbol: toe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079317.3 | NP_524041.2 | 5 - 6 | 1640 | 62 | |
| GenBank mRNA | AY069177.1 | - | 5 - 6 | 1640 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

toe| Gene Symbol: toe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079317.3 | NP_524041.2 | 5 - 6 | 1640 | 62 | |
| GenBank mRNA | AY069177.1 | - | 5 - 6 | 1640 | 62 | |
Set Membership: |
|