Assay Details
| Gene Symbol: eIF2beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079319.4 | NP_524043.1 | 2 - 3 | 548 | 62 | |
| GenBank mRNA | BT126273.1 | - | 2 - 3 | 504 | 62 | |
| L19197.1 | - | 2 - 3 | 519 | 62 | ||
More Information
Set Membership: |
|

eIF2beta
Genomic Map