Assay Details
| Gene Symbol: Tsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079320.3 | NP_524044.1 | 5 - 6 | 1346 | 55 | |
| GenBank mRNA | BT004490.1 | - | 5 - 6 | 1317 | 55 | |
More Information
Set Membership: |
|

Tsf2
Genomic Map