Assay Details
| Gene Symbol: Tom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079349.3 | NP_524073.2 | 1 | 740 | 121 | |
| GenBank mRNA | AY060362.1 | - | 1 | 722 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tom| Gene Symbol: Tom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079349.3 | NP_524073.2 | 1 | 740 | 121 | |
| GenBank mRNA | AY060362.1 | - | 1 | 722 | 121 | |
Set Membership: |
|