Assay Details
| Gene Symbol: mnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079350.5 | NP_524074.1 | 2 - 3 | 1057 | 89 | |
| NM_168600.3 | NP_730005.3 | 3 - 4 | 612 | 89 | ||
| NM_168601.3 | NP_730006.1 | 3 - 4 | 942 | 89 | ||
| GenBank mRNA | AF139834.1 | - | 2 - 3 | 1029 | 89 | |
| AY069533.1 | - | 2 - 3 | 1035 | 89 | ||
More Information
Set Membership: |
|

mnd
Genomic Map