Assay Details
| Gene Symbol: Toll-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259836.2 | NP_001246765.1 | 1 | 5259 | 109 | |
| NM_001259837.3 | NP_001246766.1 | 1 | 5259 | 109 | ||
| NM_079357.4 | NP_524081.1 | 1 | 5259 | 109 | ||
| GenBank mRNA | BT057976.1 | - | 1 | 5251 | 109 | |
More Information
Set Membership: |
|

Toll-6
Genomic Map