Assay Details
| Gene Symbol: Ran-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079358.3 | NP_524082.1 | 1 | 845 | 149 | |
| GenBank mRNA | AY070523.1 | - | 1 | 845 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ran-like| Gene Symbol: Ran-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079358.3 | NP_524082.1 | 1 | 845 | 149 | |
| GenBank mRNA | AY070523.1 | - | 1 | 845 | 149 | |
Set Membership: |
|