Assay Details
| Gene Symbol: Pka-C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079373.4 | NP_524097.2 | 5 - 6 | 2090 | 73 | |
| NM_168638.3 | NP_730083.2 | 6 - 7 | 1408 | 73 | ||
More Information
Set Membership: |
|

Pka-C3
Genomic Map