Assay Details
| Gene Symbol: nxf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275024.1 | NP_001261953.1 | 3 - 4 | 935 | 80 | |
| NM_079387.6 | NP_524111.3 | 4 - 5 | 1027 | 80 | ||
| NM_168674.4 | NP_730179.2 | 3 - 4 | 935 | 80 | ||
| GenBank mRNA | AJ312282.1 | - | 3 - 4 | 838 | 80 | |
| AY075582.1 | - | 4 - 5 | 1008 | 80 | ||
More Information
Set Membership: |
|

nxf2
Genomic Map