Assay Details
| Gene Symbol: Su(P) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079392.4 | NP_524116.2 | 1 - 2 | 842 | 60 | |
| GenBank mRNA | BT044124.1 | - | 1 - 2 | 841 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Su(P)| Gene Symbol: Su(P) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079392.4 | NP_524116.2 | 1 - 2 | 842 | 60 | |
| GenBank mRNA | BT044124.1 | - | 1 - 2 | 841 | 60 | |
Set Membership: |
|