Assay Details
| Gene Symbol: Su(P) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079392.4 | NP_524116.2 | 2 - 3 | 1243 | 63 | |
| GenBank mRNA | BT044124.1 | - | 2 - 3 | 1242 | 63 | |
More Information
Set Membership: |
|

Su(P)
Genomic Map