Assay Details
| Gene Symbol: mRpS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079410.3 | NP_524134.1 | 1 - 2 | 287 | 54 | |
| GenBank mRNA | AY071284.1 | - | 1 - 2 | 285 | 54 | |
More Information
Set Membership: |
|

mRpS26
Genomic Map