Assay Details
| Gene Symbol: mus304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079411.4 | NP_524135.3 | 3 - 4 | 2421 | 79 | |
| GenBank mRNA | AF224715.1 | - | 3 - 4 | 2399 | 79 | |
| AY118585.1 | - | 3 - 4 | 2399 | 79 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Drosophila melanogaster | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 mus304
mus304 Genomic Map
Genomic Map Back To Top
 Back To Top