Assay Details
| Gene Symbol: GNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079418.3 | NP_524142.2 | 1 - 2 | 327 | 90 | |
| GenBank mRNA | BT032835.1 | - | 1 - 2 | 318 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNBP1| Gene Symbol: GNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079418.3 | NP_524142.2 | 1 - 2 | 327 | 90 | |
| GenBank mRNA | BT032835.1 | - | 1 - 2 | 318 | 90 | |
Set Membership: |
|