Assay Details
| Gene Symbol: GNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079418.3 | NP_524142.2 | 3 - 4 | 1479 | 77 | |
| GenBank mRNA | BT032835.1 | - | 3 - 4 | 1470 | 77 | |
More Information
Set Membership: |
|

GNBP1
Genomic Map