Assay Details
| Gene Symbol: Kap-alpha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079443.2 | NP_524167.1 | 5 - 6 | 1563 | 70 | |
| GenBank mRNA | AF074957.1 | - | 4 - 5 | 1451 | 70 | |
| BT083428.1 | - | 5 - 6 | 1551 | 70 | ||
More Information
Set Membership: |
|

Kap-alpha1
Genomic Map