Assay Details
| Gene Symbol: Ac76E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259909.2 | NP_001246838.1 | 17 - 18 | 3856 | 115 | |
| NM_079449.4 | NP_524173.2 | 17 - 18 | 3856 | 115 | ||
More Information
Set Membership: |
|

Ac76E
Genomic Map