Assay Details
| Gene Symbol: sNPF-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275157.1 | NP_001262086.1 | 2 - 3 | 2159 | 95 | |
| NM_079452.3 | NP_524176.1 | 2 - 3 | 2531 | 95 | ||
| GenBank mRNA | AY051588.1 | - | 2 - 3 | 2506 | 95 | |
| AY192578.1 | - | 2 - 3 | 2771 | 95 | ||
More Information
Set Membership: |
|

sNPF-R
Genomic Map