Assay Details
| Gene Symbol: polo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014592.2 | NP_001014592.1 | 3 - 4 | 1375 | 70 | |
| NM_079455.4 | NP_524179.2 | 3 - 4 | 1768 | 70 | ||
| GenBank mRNA | AY095028.1 | - | 3 - 4 | 1411 | 70 | |
More Information
Set Membership: |
|

polo
Genomic Map