Assay Details
| Gene Symbol: alphaSnap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079456.4 | NP_524180.1 | 1 - 2 | 254 | 60 | |
| GenBank mRNA | AY069477.1 | - | 1 - 2 | 220 | 60 | |
| U09374.1 | - | 1 - 2 | 234 | 60 | ||
More Information
Set Membership: |
|

alphaSnap
Genomic Map