Assay Details
| Gene Symbol: alphaSnap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079456.4 | NP_524180.1 | 2 - 3 | 567 | 63 | |
| GenBank mRNA | AY069477.1 | - | 2 - 3 | 533 | 63 | |
| U09374.1 | - | 2 - 3 | 547 | 63 | ||
More Information
Set Membership: |
|

alphaSnap
Genomic Map