Assay Details
| Gene Symbol: Psn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275182.1 | NP_001262111.1 | 7 - 8 | 1350 | 78 | |
| NM_079460.2 | NP_524184.1 | 7 - 8 | 1507 | 78 | ||
| GenBank mRNA | AF017024.1 | - | 7 - 8 | 1336 | 78 | |
| AY061316.1 | - | 7 - 8 | 1507 | 78 | ||
| U77934.1 | - | 6 - 7 | 1280 | 78 | ||
More Information
Set Membership: |
|

Psn
Genomic Map